Learn More
Sigma Aldrich Fine Chemicals Biosciences BionicTM Buffer 10X Concen
Catalog No. 502891552
Encompass
Bionic Buffer is a unique alternative to traditional TBE (TRIS borate-EDTA) and TAE (TRIS acetate-EDTA) electrophoresis buffers. Bionic Buffer allows for higher voltage runs, extremely rapid running times, and high-resolution band separation.
Catalog No. 50-289-1552 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B61854L
Bionic Buffer is a unique alternative to traditional TBE (TRIS borate-EDTA) and TAE (TRIS acetate-EDTA) electrophoresis buffers. Bionic Buffer allows for higher voltage runs, extremely rapid running times, and high-resolution band separation.