Learn More
Chemglass Life Sciences BioNOC II Cell Culture Carrier, 50g/Bottle
SDP
Catalog No. 501532107
Encompass
Pure: 100% pure PET non-woven fabrics; regulation friendlyNon-pyrogenicNon-cytotoxicLight: High cell density (up to 1x109 cells/g)High S/V: High surface area to volume ratio; S/V = 300 cm-1High specific surface area (2, 400 cm2/g)Consistent: Novel surface treatment technology with no decay of hydrophilicityAll materials are manufactured according to cGMP guidelines. BioNOC II are carriers for the growth of animal, mammalian and insect cells. Most anchorage-dependent cells have been successfully grown in BioNOC II carriers in both serum-containing and serum-free medium. BioNOC II is made of 100% pure polyester non-woven fabrics according to cGMP guidelines. The fabric is specially surface-treated to make it hydrophilic and biocompatible. It is then processed through a cutting procedure to form 5mm wide and 10mm long strips.
Catalog No. 50-153-2107 Supplier Chemglass Life Sciences Supplier No. CLS134401
Pure: 100% pure PET non-woven fabrics; regulation friendlyNon-pyrogenicNon-cytotoxicLight: High cell density (up to 1x109 cells/g)High S/V: High surface area to volume ratio; S/V = 300 cm-1High specific surface area (2, 400 cm2/g)Consistent: Novel surface treatment technology with no decay of hydrophilicityAll materials are manufactured according to cGMP guidelines. BioNOC II are carriers for the growth of animal, mammalian and insect cells. Most anchorage-dependent cells have been successfully grown in BioNOC II carriers in both serum-containing and serum-free medium. BioNOC II is made of 100% pure polyester non-woven fabrics according to cGMP guidelines. The fabric is specially surface-treated to make it hydrophilic and biocompatible. It is then processed through a cutting procedure to form 5mm wide and 10mm long strips.