Learn More
Chemglass Life Sciences BioNOC II Cell Culture Carrier, 250g/Bottle
SDP
Catalog No. 501532108
Encompass
BioNOC II Cell Culture Carrier, 250g/Bottle. BioNOC II are carriers for the growth of animal, mammalian and insect cells. Most anchorage-dependent cells have been successfully grown in BioNOC II carriers in both serum-containing and serum-free medium. BioNOC II is made of 100% pure polyester non-woven fabrics according to cGMP guidelines. The fabric is specially surface-treated to make it hydrophilic and biocompatible. It is then processed through a cutting procedure to form 5mm wide and 10mm long strips.
Catalog No. 50-153-2108 Supplier Chemglass Life Sciences Supplier No. CLS134601
BioNOC II Cell Culture Carrier, 250g/Bottle. BioNOC II are carriers for the growth of animal, mammalian and insect cells. Most anchorage-dependent cells have been successfully grown in BioNOC II carriers in both serum-containing and serum-free medium. BioNOC II is made of 100% pure polyester non-woven fabrics according to cGMP guidelines. The fabric is specially surface-treated to make it hydrophilic and biocompatible. It is then processed through a cutting procedure to form 5mm wide and 10mm long strips.