Learn More
Advanced Biomatrix Printing probe for 12-well plates for BIONOVA X DLP Printer
Catalog No. 502255978
Encompass_Preferred
BIONOVA X Printing Probes are compatible with the BIONOVA X 3D bioprinter from CELLINK, the only DLP based bioprinter for direct in-well printing. Available for 6, 12 and 24-well multi-well printing. Each probe comes sterile and is capable of 24-hours of continuous printing.
Catalog No. 50-225-5978 Supplier Advanced Biomatrix Supplier No. D16110025292
BIONOVA X Printing Probes are compatible with the BIONOVA X 3D bioprinter from CELLINK, the only DLP based bioprinter for direct in-well printing. Available for 6, 12 and 24-well multi-well printing. Each probe comes sterile and is capable of 24-hours of continuous printing.