Learn More
Cayman Chemical BIOnT FluorIn Fragmnt lbr 10mg
SDP
Catalog No. 502943248
Encompass_Preferred
Customers are supplied with the following data package for EAch fragment purchases: • Aqueous buffer 19F NMR raw data file • Aqueous buffer 1H NMR raw data file • 19F NMR chemical shifts • Structures and physiochemical properties in an sd file The parameters used in the design of the library are: • Heavy atoms ≤ 21 • clogP ≤ 3 • Hydrogen bond donors ≤ 2 • Hydrogen bond acceptors ≤ 4 • tPSA ≤ 72 • Rotatable bonds ≤ 3
Catalog No. 50-294-3248 Supplier Cayman Chemical Supplier No. 3978710 MG
Add to Cart
Add to Cart
Request a Quote
Customers are supplied with the following data package for EAch fragment purchases: • Aqueous buffer 19F NMR raw data file • Aqueous buffer 1H NMR raw data file • 19F NMR chemical shifts • Structures and physiochemical properties in an sd file The parameters used in the design of the library are: • Heavy atoms ≤ 21 • clogP ≤ 3 • Hydrogen bond donors ≤ 2 • Hydrogen bond acceptors ≤ 4 • tPSA ≤ 72 • Rotatable bonds ≤ 3