Learn More
Cayman Chemical BIOnT PROTAC FRGmnt lbrar 10mg
SDP
Catalog No. 502942205
Encompass_Preferred
Customers are supplied with the following data package for EAch fragment purchases: • Aqueous buffer 1H NMR raw data file • Structures and physiochemical properties in an sd file • SPR Clean Screen results The parameters used in the design of the library are: • Heavy atoms ≤ 16 • clogP ≤ 3 • Hydrogen bond donors ≤ 1 • Hydrogen bond acceptors ≤ 3 • tPSA ≤ 60 • Rotatable bonds ≤ 3
Catalog No. 50-294-2205 Supplier Cayman Chemical Supplier No. 3979110 MG
Add to Cart
Add to Cart
Request a Quote
Customers are supplied with the following data package for EAch fragment purchases: • Aqueous buffer 1H NMR raw data file • Structures and physiochemical properties in an sd file • SPR Clean Screen results The parameters used in the design of the library are: • Heavy atoms ≤ 16 • clogP ≤ 3 • Hydrogen bond donors ≤ 1 • Hydrogen bond acceptors ≤ 3 • tPSA ≤ 60 • Rotatable bonds ≤ 3