Learn More
World Precision Instrument BioOxy Sensor 50 µm Tip ST connector for OxyMicro 2pcs
SDP
Catalog No. 502538657
Encompass
A new and innovative technology for measuring oxygen in gaseous and aqueous phase with important advantages over using common Clark type electrodes.
Catalog No. 50-253-8657 Supplier World Precision Instrument Supplier No. O2T0050L25012012
A new and innovative technology for measuring oxygen in gaseous and aqueous phase with important advantages over using common Clark type electrodes.