Learn More
AirSea USA Biopack 2 - 1.5L UN Combination Packaging, 4G/Class 6.2
Catalog No. 501534815
Encompass_Preferred
Biopack 2 combination packaging - supplied with 1 x 1.5L bioJar Polypropylene Secondary Packaging (Code 490). The biojar is fully autoclavable and is capable of withstanding 95kPa. The outer packaging has a high quality protective gloss lacquered finish and comes complete with hazard labels, absorbent cushioning and packaging instructions.
Catalog No. 50-153-4815 Supplier AirSea USA Supplier No. 500
Biopack 2 combination packaging - supplied with 1 x 1.5L bioJar Polypropylene Secondary Packaging (Code 490). The biojar is fully autoclavable and is capable of withstanding 95kPa. The outer packaging has a high quality protective gloss lacquered finish and comes complete with hazard labels, absorbent cushioning and packaging instructions.