Learn More
LaMotte™ BioPaddle™ Total Coliform Activity
SDPSCI_ED
Catalog No. S04348 Shop All LaMotte Company Products
Change view
Click to view available options
Agar paddle; Nutrient-TTC/MacConkey
Catalog No.
S04348 Agar paddle; Nutrient-TTC/MacConkey
1 options
Catalog No. S04348 Supplier LaMotte™ Supplier No. 5846
Includes
  • Ten paddles
  • Downloadable resources - Teacher and Student
  • Teacher and student review PowerPoint™ and iPad™/iPod™-formatted material that provides helpful background information on coliforms, field testing, Total Viable Count (TVC) and Total Coliform Count (TCC) enumeration
  • Free BioPaddle Colony Identification app
For fast and reliable cultivation and enumeration of coliform bacteria.
  • Contains BioPaddles with Nutrient-TTC/MacConkey Agar for field sampling cultivation and enumeration of coliform bacteria Total Coliform Count (TCC)
  • Gram (-) bacteria colonies appear as red dots; Gram (+) bacteria and Gram (+) cocci are usually inhibited
  • Medium gives improved differentiation between coliforms and nonlactose fermenting organisms
Order Info
Ships on Monday and Tuesday ONLY. Additional TTC/MacConkey Agar can be purchased separately.
Specifications
Manufacturing Origin United States
Media Type Nutrient-TTC/MacConkey Agar
For Use With (Application) For field sampling cultivation, enumeration of Coliform bacteria, differentiation between Coliforms and nonlactose fermenting organisms
Includes Ten paddles
Packaging Type Paddle
Quantity Each