Learn More
LaMotte™ BioPaddles™ Dual-Agar Paddles for Environmental Education

Shop All LaMotte Company Products


Description
Includes
Free basic microbe identification app and access to technical information sheet
Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.
- Longer shelf life than traditional petri dishes
- No refrigeration required
- No need for inoculating loops or Bunsen burners
- Requires simple incubation and a magnifier
- Unique design enhances colony morphology characterization
- Ten paddles per box
- Nutrient Agar: For routine culture of non-fastidious bacteria
- Sabouraud Dextrose Agar: For selective cultivation of fungi (yeasts and molds)
- Tryptic Soy (TSA)/Rose Bengal (RB) Agar: For cultivation of a wide variety of microorganisms (TSA) and selective isolation of yeasts and molds (RB)
- Nutrient TTC/MacConkey: For field sampling cultivation and enumeration of coliform bacteria total coliform count, Gram (-) bacterial colonies appear as red dots, Gram (+) bacteria are usually inhibited. MAC: Medium gives improved differentiation between coliforms and nonlactose fermenting organisms; Gram (+) cocci are usually inhibited
Order Info
Ships on Monday and Tuesday ONLY.
Specifications
Specifications
| Color | Offwhite |
| Content And Storage | 18°C to 25°C |
| Manufacturing Origin | United States |
| Media Type | TSA/RB dual Agar |
| Sterility | Sterile |
| For Use With (Application) | Microbial culture, enumeration |
| Includes | Free basic microbe identification application and access to technical information sheet |
| Packaging Type | Paddle |
| Quantity | 40 mL |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.