Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ BioPaddles™ Dual-Agar Paddles for Environmental Education
SDPSCI_ED

Catalog No. S04347 Shop All LaMotte Company Products
Change view
Click to view available options
For Use With (Application):
Bacterial culture
Bacterial isolation, testing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S04347 Bacterial isolation, testing
S04344 Bacterial culture
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04347 Supplier LaMotte™ Supplier No. 5553

missing translation for 'validMainframeMsg'

Includes

Free basic microbe identification app and access to technical information sheet

Contain microbe-specific media enclosed in a sterile vial.

Flexible, ready-to-use paddles identify and quantify microbes in air, soil, water or on any surface.

  • Longer shelf life than traditional petri dishes
  • No refrigeration required
  • No need for inoculating loops or Bunsen burners
  • Requires simple incubation and a magnifier
  • Unique design enhances colony morphology characterization
  • Ten paddles per box
  • Nutrient Agar: For routine culture of non-fastidious bacteria
  • Sabouraud Dextrose Agar: For selective cultivation of fungi (yeasts and molds)
  • Tryptic Soy (TSA)/Rose Bengal (RB) Agar: For cultivation of a wide variety of microorganisms (TSA) and selective isolation of yeasts and molds (RB)
  • Nutrient TTC/MacConkey: For field sampling cultivation and enumeration of coliform bacteria total coliform count, Gram (-) bacterial colonies appear as red dots, Gram (+) bacteria are usually inhibited. MAC: Medium gives improved differentiation between coliforms and nonlactose fermenting organisms; Gram (+) cocci are usually inhibited

Order Info

Ships on Monday and Tuesday ONLY.

Specifications

Color Yellow
Content And Storage 18°C to 25°C
Manufacturing Origin United States
Media Type NUT-TTC/MAC dual Agar
Sterility Sterile
For Use With (Application) Bacterial isolation, testing
Includes Free basic microbe identification application and access to technical information sheet
Packaging Type Paddle
Quantity 40 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.