Learn More
Med Vet International Miltex Biopblade Shave Biopsy Blade, Sterile, 50/Box
Catalog No. 502829430
Encompass_Preferred
The Miltex 33-100 Shave Biopblade is a sterile, flexible biopsy blade and is packaged 50/box. The blades are made of stainless steel. Features: Incorporates a super sharp blade with rear finger guard Ideal for tissue saucerization and shave biopsy Flexible design allows the blade to be positioned at any angle for the intended procedure Disposable, one-time use only Sterile, 5-year shelf life
Catalog No. 50-282-9430 Supplier Med Vet International Supplier No. MIL33100
The Miltex 33-100 Shave Biopblade is a sterile, flexible biopsy blade and is packaged 50/box. The blades are made of stainless steel. Features: Incorporates a super sharp blade with rear finger guard Ideal for tissue saucerization and shave biopsy Flexible design allows the blade to be positioned at any angle for the intended procedure Disposable, one-time use only Sterile, 5-year shelf life