Learn More
Labnet International, Inc BioPette Plus, 8 Channels, 20-200 uL
Catalog No. NC0974677
Encompass_Preferred
Ideal for all standard 96 well formats, the BioPette Plus Multichannel Pipettors stand up to the wear and tear of constant use while maintaining excellent accuracy and reproducibility. Four overlapping volume ranges are available to precisely meet liquid handling requirements from 1 uL to 300 uL.
Catalog No. NC0974677 Supplier Labnet International, Inc Supplier No. P4808200
Ideal for all standard 96 well formats, the BioPette Plus Multichannel Pipettors stand up to the wear and tear of constant use while maintaining excellent accuracy and reproducibility. Four overlapping volume ranges are available to precisely meet liquid handling requirements from 1 uL to 300 uL.