Learn More
Braintree Scientific BioPette Plus Autoclavable Pipette, 8-Channel,20 - 200ul
SDP
Supplier:  Braintree Scientific P4808200
Encompass
BioPette Plus Autoclavable Pipette, 8-Channel Pipette - Autoclavable pipettes designed for exceptional comfort and accuracy. Lightweight and well balanced, they fit comfortably in either right or left hand with large digital display visible to user. Chemically resistant, maintenance-free seals and finely polished pistons provide unsurpassed accuracy sample after sample. Continuously adjustable, digital micrometer is slightly recessed to prevent unintentional change and is protected from effects of thermal expansion by insulated body of the pipette. For convenience, volume can also be adjusted by turning push-button. Ergonomically positioned, tip ejector reduces fatigue during prolonged use. Discovery pipettes have a universal shaft that accepts most pipette tips. Tip ejector is adjustable to accommodate various styles of tips and removable for pipetting into narrow tubes. Made from stainless steel, it will not rust, corrode or break. Discovery pipettes are fully autoclavable.
Catalog No. 50-195-5466