Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Spooled Precision Pump Tubing, BioPharm Platinum-Cured Silicone, L/S 14; 400 ft
Catalog No. NC9732696
Encompass
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
Catalog No. NC9732696 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9642314
Applications: Pharmaceutical, cosmetic, research, and auto-analysis applications Characteristics: Ultra-smooth, minimizes bacterial growth, good for mild to medium concentration bases, salts, and alcohols; odorless, tasteless, nontoxic, transparent Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.