Learn More
Waters Corp Training Kit, BioPharma, Peptide, Standards
Catalog No. 502392924 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This training kit provides an array of compounds for education, demonstration, and system applications in the biopharmaceutical industry.
  • Contains 1 vial of 2.5 mg horseheart myoglobin.
  • Contains 1 vial of 1 mg intact mAb check standard.
  • Contains 1 vial of 0.5 mg Glu-fibrinopeptideB.
  • Contains 1 vial of 1 nmol MassPrep enolase digest.
  • Contains 1 vial of 1 mg each reverse peptide Ser-Asp-Gyl-Arg-Gyl.
  • Contains 1 vial of 1 mg each reverse peptide Gly-Arg-Gyl-Asp-Ser.
  • Supports peptide applications.
Catalog No. 50-239-2924 Supplier Waters Corp Supplier No. 186006921
This training kit provides an array of compounds for education, demonstration, and system applications in the biopharmaceutical industry.
  • Contains 1 vial of 2.5 mg horseheart myoglobin.
  • Contains 1 vial of 1 mg intact mAb check standard.
  • Contains 1 vial of 0.5 mg Glu-fibrinopeptideB.
  • Contains 1 vial of 1 nmol MassPrep enolase digest.
  • Contains 1 vial of 1 mg each reverse peptide Ser-Asp-Gyl-Arg-Gyl.
  • Contains 1 vial of 1 mg each reverse peptide Gly-Arg-Gyl-Asp-Ser.
  • Supports peptide applications.