Learn More
Waters Corp Biopharmaceutical Preparation Kit, for Training Standards, MS Detection
Catalog No. 502392787 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit enables the preparation of Biopharmaceutical Training Standards. It contains a 1 to 2 mL ampule of formic acid, a 1 to 30 mL Nalgene container of 2μg/μL sodium iodide, 50 ng/μL cesium iodide in 50:50 Isopropanol:water, and three 30 mL empty fluidic bottles with caps. Care and Use instructions are available in document 720004859en.
  • Enables preparation of biopharmaceutical training standards.
  • Includes a 1 to 2 mL ampule of formic acid.
  • Includes a 1 to 30 mL Nalgene container of 2 μg/μL sodium iodide.
  • Includes 50 ng/μL cesium iodide in 50:50 isopropanol:water.
  • Includes three 30 mL empty fluidic bottles with caps.
  • Supports detection methods such as MS, MS/MS, UV, and HRMS.
  • Designed for protein application.
Catalog No. 50-239-2787 Supplier Waters Corp Supplier No. 186007517
Add to Cart
Add to Cart
This kit enables the preparation of Biopharmaceutical Training Standards. It contains a 1 to 2 mL ampule of formic acid, a 1 to 30 mL Nalgene container of 2μg/μL sodium iodide, 50 ng/μL cesium iodide in 50:50 Isopropanol:water, and three 30 mL empty fluidic bottles with caps. Care and Use instructions are available in document 720004859en.
  • Enables preparation of biopharmaceutical training standards.
  • Includes a 1 to 2 mL ampule of formic acid.
  • Includes a 1 to 30 mL Nalgene container of 2 μg/μL sodium iodide.
  • Includes 50 ng/μL cesium iodide in 50:50 isopropanol:water.
  • Includes three 30 mL empty fluidic bottles with caps.
  • Supports detection methods such as MS, MS/MS, UV, and HRMS.
  • Designed for protein application.