Learn More
AB Sciex LLC BIOPHASE CIEF KIT
Catalog No. NC3366289
Encompass
  • Configured for high-resolution separation of proteins based on pI
  • Compatible with the BioPhase 8800 system
  • Supports quantitative analysis of charge variants
  • Comprehensive kit contains reagents for 200 cIEF tests
  • Ideal for protein characterization and pI determination
  • Includes anodic stabilizer, anolyte, cathodic stabilizer, catholyte, CE grade water, cIEF gel, chemical mobilizer, formamide, neutral capillary conditioning solution, urea
  • Target analytes include proteins and their charge variants
  • For research use only
Catalog No. NC3366289 Supplier AB Sciex LLC Supplier No. C30101
  • Configured for high-resolution separation of proteins based on pI
  • Compatible with the BioPhase 8800 system
  • Supports quantitative analysis of charge variants
  • Comprehensive kit contains reagents for 200 cIEF tests
  • Ideal for protein characterization and pI determination
  • Includes anodic stabilizer, anolyte, cathodic stabilizer, catholyte, CE grade water, cIEF gel, chemical mobilizer, formamide, neutral capillary conditioning solution, urea
  • Target analytes include proteins and their charge variants
  • For research use only