Learn More
AB Sciex LLC BIOPHASE NEUT CARTRIDGE 8X30CM
Catalog No. NC3357876
Encompass
  • High-resolution separation of complex biomolecules
  • 8 capillaries with a length of 30 cm
  • Neutral coating for effective capillary electrophoresis
  • Compatible with the BioPhase 8800 system
  • Integrated liquid temperature control for standardized separation temperatures
  • High assay repeatability for consistent results
  • Incorporated electrodes for enhanced detection
  • Detection window located 20 cm from inlet
  • Designed for analyzing proteins and nucleic acids
  • For research use only
  • Ships in a storage box for protection
Catalog No. NC3357876 Supplier AB Sciex LLC Supplier No. 5080119
  • High-resolution separation of complex biomolecules
  • 8 capillaries with a length of 30 cm
  • Neutral coating for effective capillary electrophoresis
  • Compatible with the BioPhase 8800 system
  • Integrated liquid temperature control for standardized separation temperatures
  • High assay repeatability for consistent results
  • Incorporated electrodes for enhanced detection
  • Detection window located 20 cm from inlet
  • Designed for analyzing proteins and nucleic acids
  • For research use only
  • Ships in a storage box for protection