Learn More
Aniara Diagnostica LLC BIOPHEN CS-01(38) THROMB 12 EA
Supplier:  Aniara Diagnostica LLC A229001C
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Chromogenic substrate for thrombin, lyophilized. Peptide sequence H-D-Phe-Pip-Arg-pNa, 2HCl. 12 vials x 25 mg.
Catalog No. 50-100-8926
Add to Cart
Request a Quote