Learn More
Aniara Diagnostica LLC BIOPHEN CS-01(38) THROMBIN
Catalog No. 501008925
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Chromogenic substrate for thrombin, lyophilized. Peptide sequence H-D-Phe-Pip-Arg-pNa, 2HCl. 1 vial x 25 mg.
Catalog No. 50-100-8925 Supplier Aniara Diagnostica LLC Supplier No. A229001
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Chromogenic substrate for thrombin, lyophilized. Peptide sequence H-D-Phe-Pip-Arg-pNa, 2HCl. 1 vial x 25 mg.