Learn More
Aniara Diagnostica LLC BIOPHEN CS-11(65) FXA
Catalog No. 501008928
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Chromogenic substrate for Factor Xa, lyophilized. Peptide sequence D-Arg-Gly-Arg-pNA, 2HCl. 1 vial x 25 mg.
Catalog No. 50-100-8928 Supplier Aniara Diagnostica LLC Supplier No. A229014
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Chromogenic substrate for Factor Xa, lyophilized. Peptide sequence D-Arg-Gly-Arg-pNA, 2HCl. 1 vial x 25 mg.