Learn More
Aniara Diagnostica LLC BIOPHEN FACTOR IX (6) ASSAY
Catalog No. 501008889
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor IX (6) Assay Kit. Quantitative chromogenic assay for Factor IX activity in human citrated plasma, biological fluids, or concentrates. Manual Microplate (2x120 Tests) or Automated.
Catalog No. 50-100-8889 Supplier Aniara Diagnostica LLC Supplier No. A221806RUO
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor IX (6) Assay Kit. Quantitative chromogenic assay for Factor IX activity in human citrated plasma, biological fluids, or concentrates. Manual Microplate (2x120 Tests) or Automated.