Learn More
Aniara Diagnostica LLC BIOPHEN FACTOR VII ASSAY
Catalog No. 501008884
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor VII Assay Kit. Quantitative chromogenic assay for Factor VII activity in human citrated plasma or biological fluids. Manual Microplate (2x65 Tests) or Automated.
Catalog No. 50-100-8884 Supplier Aniara Diagnostica LLC Supplier No. A221304
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor VII Assay Kit. Quantitative chromogenic assay for Factor VII activity in human citrated plasma or biological fluids. Manual Microplate (2x65 Tests) or Automated.