Learn More
Aniara Diagnostica LLC BIOPHEN FVIII:C (6) ASSAY
Catalog No. 501008886
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor VIII:C (6) Assay Kit. Quantitative chromogenic assay for Factor VIII:C activity in human citrated plasma, biological fluids, or concentrates. Manual Microplate (2x120 Tests) or Automated.
Catalog No. 50-100-8886 Supplier Aniara Diagnostica LLC Supplier No. A221406RUO
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. BIOPHEN Factor VIII:C (6) Assay Kit. Quantitative chromogenic assay for Factor VIII:C activity in human citrated plasma, biological fluids, or concentrates. Manual Microplate (2x120 Tests) or Automated.