Learn More
Aniara Diagnostica LLC BIOPHEN RIVAROXABAN CONTROL
Catalog No. 501008916
Encompass
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Normal human plasma with Rivaroxaban, lyophilized. 2 levels at about 100 and 300 ng/mL. Use with chromogenic anti-Xa assays. Concentrations established against reference preparation with BIOPHEN DiXal (A221030-RUO). 6x2x1 mL vials.
Catalog No. 50-100-8916 Supplier Aniara Diagnostica LLC Supplier No. A224501RUO
This item MFG by HYPHEN BioMed and Dis. by Aniara Diagnostics. Normal human plasma with Rivaroxaban, lyophilized. 2 levels at about 100 and 300 ng/mL. Use with chromogenic anti-Xa assays. Concentrations established against reference preparation with BIOPHEN DiXal (A221030-RUO). 6x2x1 mL vials.