Learn More
Bio Rad Laboratories BIOPLEX CALIBRATION KIT
Catalog No. NC9879994
Encompass
Bio-Plex Calibration Kit, Includes CAL1 and CAL2 Calibration Beads for Approximately 50 Daily Calibration Routines
Catalog No. NC9879994 Supplier Bio Rad Laboratories Supplier No. 171203060
Bio-Plex Calibration Kit, Includes CAL1 and CAL2 Calibration Beads for Approximately 50 Daily Calibration Routines