Learn More
YMC America, Inc BioPro QF Acc 3um 100x2.1mm
SDP
Catalog No. 5000880807
Encompass
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) The non-porous formats (QF SF) are highly efficient and are therefore ideal for high-throughput analytical separations requiring high resolution
Catalog No. 50-008-80807 Supplier YMC America, Inc Supplier No. QF00S0310Q1PTC
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) The non-porous formats (QF SF) are highly efficient and are therefore ideal for high-throughput analytical separations requiring high resolution