Learn More
YMC America, Inc BioPro SF Acc 5um 250x4.6mm
SDP
Catalog No. 5000876889
Encompass
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) The non-porous formats (QF SF) are highly efficient and are therefore ideal for high-throughput analytical separations requiring high resolution
Catalog No. 50-008-76889 Supplier YMC America, Inc Supplier No. SF00S052546PTC
Add to Cart
Add to Cart
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) The non-porous formats (QF SF) are highly efficient and are therefore ideal for high-throughput analytical separations requiring high resolution