Learn More
YMC America, Inc BioPro SP PEEK 5um 150x4.6mm
SDP
Catalog No. 5000880612
Encompass
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) Columns packed with the porous materials (QA SP) are advantageous for high-load analytical separations and laboratory-scale purification as they maintain strong peak shape even with increased load
Catalog No. 50-008-80612 Supplier YMC America, Inc Supplier No. SPA0S051546WP
YMC-BioPro products are made with a hydrophilic polymer base particle and are used for the separation of biomolecules This column family delivers reliable high-resolution separations from analytical to preparative scale YMC-BioPro IEX columns are available with porous and non-porous particles for strong anion exchange (SAX) and strong cation exchange (SCX) Columns packed with the porous materials (QA SP) are advantageous for high-load analytical separations and laboratory-scale purification as they maintain strong peak shape even with increased load