Learn More
Thermo Scientific™ BioProcess Container Support Tote
GREENER_CHOICE
Catalog No. SV3020001 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
10 to 20 L
50 mL to 5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SV3020001 50 mL to 5 L
SV3020002 10 to 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SV3020001 Supplier Thermo Scientific™ Supplier No. SV30200.01
Add to Cart
Add to Cart
The Thermo Scientific BioProcess Container (BPC) Support Tote was designed to meet the demands of various bioprocessing workflow requirements.
The Thermo Scientific BioProcess Container (BPC) Support Tote was designed to meet the demands of various bioprocessing workflow requirements. Made of high-grade high-density polyethylene (HDPE), the tote is exceptionally durable and chemical-resistant. The tote is available in two sizes: the small tote is for Thermo Scientific BPCs up to 5 L, and the large tote is for 10-L and 20-L BPCs. Both sizes have tubing access windows that allow for filling, draining, and sampling while providing protection from light.
Features of the BPC Support Tote include:
  • Light protection—the lid offers protection for light-sensitive products while providing access to BPC for in-process use
  • Optimized drainage—a kickstand on the large tote enables you to slope the tote for improved drainage; smaller tote features built-in sloped design
  • Validated stackable—totes can be stacked with lids attached, to decrease the overall footprint
  • Secondary containment—the recessed floor of the tote provides secondary containment in the unlikely event of a leak
  • Nestable—totes rest inside one another to minimize storage space needed for processes that require multiple totes
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL to 5 L
Quantity 1 tote
Shipping Condition Room Temperature
Product Line Thermo Scientific