Learn More
Nova Biomedical Corporation Nova Biomedical - 58117 - BioProfile CDV V2 Reagent Pack0
Catalog No. NC1170603
Encompass
BioProfile CDV V2 Reagent Pack- BioProfile CDV incorporates advanced, state-of-the-art technology for rapid, accurate measurement of cell density and cell viability in cell cultures. Based on the widely recognized and accepted trypan blue exclusion method, BioProfile CDV combines advanced technology in robotics, optics and computer algorithms in a highly automated analyzer.
Catalog No. NC1170603 Supplier Nova Biomedical Corporation Supplier No. 58117
BioProfile CDV V2 Reagent Pack- BioProfile CDV incorporates advanced, state-of-the-art technology for rapid, accurate measurement of cell density and cell viability in cell cultures. Based on the widely recognized and accepted trypan blue exclusion method, BioProfile CDV combines advanced technology in robotics, optics and computer algorithms in a highly automated analyzer.