Learn More
YMC America, Inc BioProSS Q20 20um 150x30mm
SDP
Catalog No. 5000881840
Encompass
YMC s BioPro IEX resins are fully scalable exhibiting consistent chromatographic behavior across all particle sizes YMC-BioPro IEX SmartSep Q/S (10 20 or 30 um) which show high resolution and recovery even at a high flow rate and high loading condition are suitable for an intermediate purification step and a polishing step of mAbs proteins peptides and oligonucleotides
Catalog No. 50-008-81840 Supplier YMC America, Inc Supplier No. QSA0S201530WT
YMC s BioPro IEX resins are fully scalable exhibiting consistent chromatographic behavior across all particle sizes YMC-BioPro IEX SmartSep Q/S (10 20 or 30 um) which show high resolution and recovery even at a high flow rate and high loading condition are suitable for an intermediate purification step and a polishing step of mAbs proteins peptides and oligonucleotides