Learn More
Leica Biosystems BIOPSY BAG SM 30INX45MM 500/PK
Catalog No. NC1213450
Encompass_Preferred
Manufactured from solvent resistant nylon, these Mesh Biopsy Bags are ideal for use in processing small biopsies. They provide fluid exchange and tissue safety. The fixative and specimen can be poured into the bag which then filters the fixative and leaves the biopsies inside. The bag is then closed, placed inside a Biopsy Cassette and processed as normal. After processing, the bag can be extracted and opened, allowing the specimen to be recovered and embedded in paraffin wax.
This item is not returnable. View return policy
Catalog No. NC1213450 Supplier Leica Biosystems Supplier No. 3801085
Manufactured from solvent resistant nylon, these Mesh Biopsy Bags are ideal for use in processing small biopsies. They provide fluid exchange and tissue safety. The fixative and specimen can be poured into the bag which then filters the fixative and leaves the biopsies inside. The bag is then closed, placed inside a Biopsy Cassette and processed as normal. After processing, the bag can be extracted and opened, allowing the specimen to be recovered and embedded in paraffin wax.