Learn More
IMEB, Inc Tek-Select Type 8 Biopsy Cassettes with Hinged Lid, Yellow, 1000 per Case, Frosted Easy to Write On, Lids Attached
SDP
Catalog No. 501689514
Encompass_Preferred
Tek-Select Type 8 Biopsy Cassettes offer a special hinge holding the base and cover together. This hinge allows cassettes to be opened and closed as often as necessary. The lid can be removed and re-inserted easily without danger of specimen loss. Three frosted writing surfaces are easy to write on, including the 45 deg. angle anterior face. Cassettes are packaged 1000 per case, with lids attached. Our best selling biopsy cassettes.
Catalog No. 50-168-9514 Supplier IMEB, Inc Supplier No. CY881
Tek-Select Type 8 Biopsy Cassettes offer a special hinge holding the base and cover together. This hinge allows cassettes to be opened and closed as often as necessary. The lid can be removed and re-inserted easily without danger of specimen loss. Three frosted writing surfaces are easy to write on, including the 45 deg. angle anterior face. Cassettes are packaged 1000 per case, with lids attached. Our best selling biopsy cassettes.