Learn More
Azer Scientific Biopsy Cassettes, Lids Packaged Seperately, Taped
SDP
Catalog No. 22026829
Change view
Click to view available options
Color:
Blue
Green
Lilac
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-026-829 White
22-026-821 Blue
22-026-822 Green
22-026-824 Lilac
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-026-829 Supplier Azer Scientific Supplier No. ES6048W
Add to Cart
Add to Cart
This item is not returnable. View return policy
Biopsy cassettes, lids packaged separately, taped, 40/stack, 50 stacks/case.
These cassettes are specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets. Molded from acetal, they keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage.
This model differs by the special hinge that holds the base and cover together. The hinge design configuration allows for multiple opening and closing of the lid.
Features:
  • Made of acetal
  • 1 mm square openings to maximize fluid exchange
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
  • Cassettes and lids packaged separately
Specifications
Closure Included Yes
Closure Type Lid
Color White
Product Type Biopsy Cassette