Learn More
Electron Microscopy Sciences Multi-compartment Biopsy Cassette 4-compartment
SDP
Catalog No. 501931164
Encompass_Preferred
The new EMS multi-compartment biopsy cassettes are available with 2, 4 or 9 compartments with 0.6mm holes for added efficiency. Made from high density acetal they are resistant to histological solvents like alcohol and xylene. Casettes are preassembled with easy to open and close lids.
This item is not returnable. View return policy
Catalog No. 50-193-1164 Supplier Electron Microscopy Sciences Supplier No. 7006102
The new EMS multi-compartment biopsy cassettes are available with 2, 4 or 9 compartments with 0.6mm holes for added efficiency. Made from high density acetal they are resistant to histological solvents like alcohol and xylene. Casettes are preassembled with easy to open and close lids.