Learn More
Electron Microscopy Sciences IP ActivFlo Biopsy I Cassettes, Taped Tan
SDP
Catalog No. 501926984
Encompass_Preferred
Patented "Fluidic Flow" pore design maximizes reagent exchange. IP ActivFlo Biopsy Cassettes feature innovative V-shaped 0.67 mm pores that pull fluid in and force air out. Sloped walls create a basket for small tissue management. The one-piece cassettes with 'frangible lids' have been designed for the Leica IPC Cassette Printer, with <1% printing error rate.
Color: Taped Tan
This item is not returnable. View return policy
Catalog No. 50-192-6984 Supplier Electron Microscopy Sciences Supplier No. 7007629
Patented "Fluidic Flow" pore design maximizes reagent exchange. IP ActivFlo Biopsy Cassettes feature innovative V-shaped 0.67 mm pores that pull fluid in and force air out. Sloped walls create a basket for small tissue management. The one-piece cassettes with 'frangible lids' have been designed for the Leica IPC Cassette Printer, with <1% printing error rate.
Color: Taped Tan