Learn More
Electron Microscopy Sciences Slimsette™ Biopsy Cassettes in QuickLoad™ Stacks, Pink
SDP
Catalog No. 501930808
Encompass_Preferred
Cassettes, for use with Leica and Sakura printers, are molded from a special high density acetal. They keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage. The cover does not protrude above the cassette, a great space saving feature. The anterior printing area is at a 45 degree angle and offers an unobstructed view of the writing surface.
The one-piece disposable plastic cover is pre-installed on each cassette and eliminates the need for reusable steel lids. The cover can be opened and closed as often as necessary and will always relock, reducing the possibility of specimen loss. For Leica ans Sakura printers.
Color: Pink
This item is not returnable. View return policy
Catalog No. 50-193-0808 Supplier Electron Microscopy Sciences Supplier No. 7007975
Cassettes, for use with Leica and Sakura printers, are molded from a special high density acetal. They keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximize fluid exchange and ensure proper reagent drainage. The cover does not protrude above the cassette, a great space saving feature. The anterior printing area is at a 45 degree angle and offers an unobstructed view of the writing surface.
The one-piece disposable plastic cover is pre-installed on each cassette and eliminates the need for reusable steel lids. The cover can be opened and closed as often as necessary and will always relock, reducing the possibility of specimen loss. For Leica ans Sakura printers.
Color: Pink