Learn More
Alkali Scientific Biopsy Embedding Cassette with Attached (Detachable) Lid 35° Writing Area, Dispenser Box, 500/Pk, Yellow
SDP
Catalog No. 5000662012
Encompass_Preferred
These 35° angled biopsy embedding cassettes are designed for the secure processing of small and delicate tissue samples. Precision molded fine mesh openings help prevent specimen loss while allowing optimal fluid exchange during processing and embedding. The integrated one piece construction features a hinged lid that remains attached, allowing repeated opening and closing without the risk of separation. The 35° angled design provides a large slanted writing surface that improves label visibility and works well with both manual marking and automated cassette printers. A reliable snap lock closure keeps the lid securely shut, protecting tissue samples throughout the workflow. Supplied in a convenient dispenser box for easy access, these cassettes support efficient organization in busy histology and pathology laboratories. Fine mesh construction is ideal for handling small or fragile biopsy specimens while maintaining secure containment during routine processing.
Catalog No. 50-006-62012 Supplier Alkali Scientific Supplier No. ES3014Y
These 35° angled biopsy embedding cassettes are designed for the secure processing of small and delicate tissue samples. Precision molded fine mesh openings help prevent specimen loss while allowing optimal fluid exchange during processing and embedding. The integrated one piece construction features a hinged lid that remains attached, allowing repeated opening and closing without the risk of separation. The 35° angled design provides a large slanted writing surface that improves label visibility and works well with both manual marking and automated cassette printers. A reliable snap lock closure keeps the lid securely shut, protecting tissue samples throughout the workflow. Supplied in a convenient dispenser box for easy access, these cassettes support efficient organization in busy histology and pathology laboratories. Fine mesh construction is ideal for handling small or fragile biopsy specimens while maintaining secure containment during routine processing.