Learn More
StatLab Biopsy Master Papers
SDP
Catalog No. 21033474
Change view
Click to view available options
Quantity:
500 Pk.
Catalog No. Quantity
21-033-474 500 Pk.
1 options
Catalog No. 21-033-474 Supplier StatLab Supplier No. TCZ03500P
Add to Cart
Add to Cart
Microporous mesh paper that prevents specimen loss during tissue processing
  • Specially developed micro-porous mesh paper that prevents specimen loss of small biopsies during tissue processing and embedding procedures
  • Each mesh paper allows uncompromised solvent exchange for complete penetration and infiltration
Specifications
Product Type Biopsy Paper
Quantity 500 Pk.