Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Biopsy NBF Kit
SDP

Catalog No. PBKNB2120CA
Change view
Click to view available options
Capacity (Metric):
2 x 120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
PBKNB2120CA 2 x 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. PBKNB2120CA Supplier StatLab Supplier No. PBKNB2/120/CASS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or if inhaled

Kit is pre-labeled with location diagram. Kit contains two 120 mL prefilled 10% NBF containers with biopsy location stickers and 12 pre-labeled cassettes.

  • NBF Features a pH level of 7.0

Specifications

Product Type Biopsy Container
Closure Included Yes
Graduated No
Material HDPE
Shape Straight, Cylindrical
Sterility Non-sterile
Autoclavable Not Autoclavable
Capacity (Metric) 2 x 120 mL
Includes Two 120 mL Prefilled Vials with 12 cassettes
Fit for Purpose Certified Quality

Prefilled container

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.