Learn More
Roboz Surgical BIOPSY NEEDLE 14 X 70MM
SDP
Catalog No. 5000574483
Encompass
This biopsy needle with stylet is a high-quality surgical instrument designed for precision sampling in laboratory and research applications. Constructed from durable, autoclavable stainless steel, it is engineered for repeated use in non-clinical settings.
  • Manufactured from high-grade stainless steel for longevity and corrosion resistance
  • Fully autoclavable design allows for repeated sterilization and reuse
  • Stylet included for effective sample retrieval and needle clearing
  • Suitable for laboratory research and experimental procedures
  • Not intended for use in human medical procedures
Catalog No. 50-005-74483 Supplier Roboz Surgical Supplier No. RS634014
This biopsy needle with stylet is a high-quality surgical instrument designed for precision sampling in laboratory and research applications. Constructed from durable, autoclavable stainless steel, it is engineered for repeated use in non-clinical settings.
  • Manufactured from high-grade stainless steel for longevity and corrosion resistance
  • Fully autoclavable design allows for repeated sterilization and reuse
  • Stylet included for effective sample retrieval and needle clearing
  • Suitable for laboratory research and experimental procedures
  • Not intended for use in human medical procedures