Learn More
Leica Biosystems Biopsy Pads, 25 x 32 mm (1 x 1.25 in) - For use with standard biopsy cassettes
Catalog No. NC3100213
Encompass
Biopsy Pads are thin, open-celled foam pads sized 25 x 32 mm (approx. 1 x 1.25 in) designed to secure small tissue specimens in standard biopsy cassettes during fixation and processing. Presaturating the pads in fixative reduces specimen floating and artifacts, while the thin construction minimizes pressure on unfixed tissue and reduces the risk of tissue sticking or fiber separation during embedding. Supplied in packs of 500.
  • Thin open-celled foam that permits thorough fluid transfer around specimens.
  • Presaturate in fixative to prevent specimen floating and reduce artifacts.
  • Thin construction minimizes pressure on unfixed tissue.
  • Reduces the risk of tissue sticking and fiber separation during embedding.
  • Sized to fit standard 1 x 1.25 in biopsy cassettes.
  • Supplied in a pack of 500 pads for high-volume use.
Catalog No. NC3100213 Supplier Leica Biosystems Supplier No. 3801020
Biopsy Pads are thin, open-celled foam pads sized 25 x 32 mm (approx. 1 x 1.25 in) designed to secure small tissue specimens in standard biopsy cassettes during fixation and processing. Presaturating the pads in fixative reduces specimen floating and artifacts, while the thin construction minimizes pressure on unfixed tissue and reduces the risk of tissue sticking or fiber separation during embedding. Supplied in packs of 500.
  • Thin open-celled foam that permits thorough fluid transfer around specimens.
  • Presaturate in fixative to prevent specimen floating and reduce artifacts.
  • Thin construction minimizes pressure on unfixed tissue.
  • Reduces the risk of tissue sticking and fiber separation during embedding.
  • Sized to fit standard 1 x 1.25 in biopsy cassettes.
  • Supplied in a pack of 500 pads for high-volume use.