Learn More
Electron Microscopy Sciences Biopsy Paper Bags, 45 mm x 75 mm, 100/pk
SDP
Catalog No. 5033339
Encompass_Preferred
These bags are made from porous paper which provides maximum tissue safety during processing. They permit proper fluid exchange and drainage. To Use simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. You are now ready to process as usual.
Catalog No. 50-333-39 Supplier Electron Microscopy Sciences Supplier No. 6232601
These bags are made from porous paper which provides maximum tissue safety during processing. They permit proper fluid exchange and drainage. To Use simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. You are now ready to process as usual.