Learn More
Electron Microscopy Sciences Biopsy Paper Filters, Black
SDP
Supplier:  Electron Microscopy Sciences 6232692
Encompass_Preferred
Specifically designed for processing biopsy specimens. Non-absorbent mesh allows fluids to flow through freely while containing small tissue fragments. Non-absorbent porous mesh prevents reagent carryover. They eliminate the indentation pattern left by form biopsy pads. Can be easily opened with tweezers. Allows efficient solvent flow-through during processing.
Color: Black
Catalog No. 50-192-8741