Learn More
Electron Microscopy Sciences Biopsy Paper Bags, 30 x 50 mm
SDP
Catalog No. 501927357
Encompass_Preferred
These bags are made from porous paper which provides maximum tissue safety during processing. They permit proper fluid exchange and drainage. To Use simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. You are now ready to process as usual.
Size: 30 x 50 mm
Catalog No. 50-192-7357 Supplier Electron Microscopy Sciences Supplier No. 6232610
These bags are made from porous paper which provides maximum tissue safety during processing. They permit proper fluid exchange and drainage. To Use simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. You are now ready to process as usual.
Size: 30 x 50 mm