Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Biopsy Punch

Catalog No. 21124665 Shop All Surgical Design Products
Change view
Click to view available options
Size:
1 mm
1.5 mm
2 mm
2.5 mm
3 mm
3.5 mm
4 mm
5 mm
6 mm
8 mm
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
21-124-665 1 mm
21-124-666 1.5 mm
21-124-667 2 mm
21-124-668 2.5 mm
21-124-669 3 mm
21-124-670 3.5 mm
21-124-671 4 mm
21-124-672 5 mm
21-124-673 6 mm
21-124-674 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 21-124-665 Supplier Surgical Design™ Supplier No. BP10BX
Add to Cart
Add to Cart

Sterile disposable biopsy punches provide precise skin and tissue sampling for medical, clinical, and histology applications.

  • Sterile, single-use biopsy punches packaged 10 per box
  • Ultra-sharp stainless steel cutting edge for clean, consistent tissue retrieval
  • Color-coded handles for quick size identification
  • Designed for dermatology, histology, pathology, and general clinical biopsy procedures
  • Produces uniform cylindrical tissue samples ideal for microtomy and microscopic evaluation
  • Ergonomic, non-slip grip for controlled tissue punching
  • Sizes available (mm): 1, 1.5, 2, 2.5, 3, 3.5, 4, 5, 6, 8
  • Low-trauma penetration for patient comfort and reduced tissue distortion
  • Individually sealed to maintain sterility until point of use
  • Latex-free, medical-grade construction
  • Suitable for skin lesions, nevi, tumors, wound margins, and soft-tissue sampling

Specifications

Product Type Biopsy Punch
For Use With (Application) Single-use Disposability Without Sacrificing Quality
Size 1 mm
Disposable Single-use
Sterility Sterile
Graduated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.