Learn More
Med Vet International 3mm Biopsy Punch, Sterile, Each
Catalog No. 502522437
Encompass_Preferred
Ergonomically designed for routine dermal proceduresFeature a unique, easy-grip barrel and pressure assistance platform for superb controlRazor-sharp stainless steel blade for precise excisions
Catalog No. 50-252-2437 Supplier Med Vet International Supplier No. BP3MM
Ergonomically designed for routine dermal proceduresFeature a unique, easy-grip barrel and pressure assistance platform for superb controlRazor-sharp stainless steel blade for precise excisions