Learn More
Med Vet International 5mm Biopsy Punch, Sterile, 10/box
Supplier:  Med Vet International BP5MMX10
Encompass_Preferred
Ergonomically designed for routine dermal proceduresFeature a unique, easy-grip barrel and pressure assistance platform for superb controlRazor-sharp stainless steel blade for precise excisions
Catalog No. 50-304-6787